accessibility menu, dialog, popup

UserName

CAS RN 2424-92-2

OOOO

CAS RN 2424-92-2

IUPAC Name: icosanedioate
Synonyms: icosanedioate
Molecular Weight (g/mol): 340.51
Molecular Formula: C20H36O4
InChi Key: JJOJFIHJIRWASH-UHFFFAOYSA-L
SMILES: [O-]C(=O)CCCCCCCCCCCCCCCCCCC([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)