accessibility menu, dialog, popup

UserName

CAS RN 2426-87-1

IUPAC Name: 4-(benzyloxy)-3-methoxybenzaldehyde
Synonyms: 4-(benzyloxy)-3-methoxybenzaldehyde
Molecular Weight (g/mol): 242.27
Molecular Formula: C15H14O3
InChi Key: JSHLOPGSDZTEGQ-UHFFFAOYSA-N
SMILES: COC1=CC(C=O)=CC=C1OCC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)