CAS RN 2443-58-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Hydroxy Fluorene, TRC
CAS: 2443-58-5 Molecular Formula: C13 H10 O Molecular Weight (g/mol): 182.22 Synonym: 9H-Fluoren-2-ol,Fluoren-2-ol (6CI,7CI,8CI),2-Hydroxy-9H-fluorene,2-Hydroxyfluorene,NSC 26316,NSC 31248 IUPAC Name: 9H-fluoren-2-ol SMILES: Oc1ccc2c(Cc3ccccc23)c1