accessibility menu, dialog, popup

UserName

CH3OOH3CFFF(E)

CAS RN 24490-03-7

IUPAC Name: ethyl (2E)-4,4,4-trifluoro-3-methylbut-2-enoate
Synonyms: ethyl (2E)-4,4,4-trifluoro-3-methylbut-2-enoate
Molecular Weight (g/mol): 182.14
Molecular Formula: C7H9F3O2
InChi Key: OSZLARYVWBUKTG-SNAWJCMRSA-N
SMILES: CCOC(=O)\C=C(/C)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results