accessibility menu, dialog, popup

UserName

CAS RN 24686-78-0

ONO

CAS RN 24686-78-0

IUPAC Name: 1-benzoylpiperidin-4-one
Synonyms: 1-benzoylpiperidin-4-one
Molecular Weight (g/mol): 203.24
Molecular Formula: C12H13NO2
InChi Key: NZAXGZYPZGEVBD-UHFFFAOYSA-N
SMILES: O=C(N1CCC(=O)CC1)C1=CC=CC=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)