CAS RN 2518276-69-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Methyl 3-(2-Ethyl-4-oxo-1-phenyl-1,3,8-triazaspiro[4.5]dec-2-en-8-yl)propanoate, TRC
CAS: 2518276-69-0 Molecular Formula: C19H25N3O3 Molecular Weight (g/mol): 343.43 Synonym: Remifentanil EP Impurity L IUPAC Name: methyl 3-(2-ethyl-4-oxo-1-phenyl-1,3,8-triazaspiro[4.5]dec-2-en-8-yl)propanoate SMILES: CCC1=NC(=O)C2(CCN(CCC(=O)OC)CC2)N1c3ccccc3