CAS RN 2518444-69-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
4-Bromo-1-(4-methoxyphenyl)-6-(4-nitrophenyl)-7-oxo-6,7-dihydro-1H-pyrazolo[3,4-c]pyridine-3-carboxylic Acid Ethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4-Bromo-1-(4-methoxyphenyl)-6-(4-nitrophenyl)-7-oxo-6,7-dihydro-1H-pyrazolo[3,4-c]pyridine-3-carboxylic Acid Ethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
4-Bromo-1-(4-methoxyphenyl)-6-(4-nitrophenyl)-7-oxo-6,7-dihydro-1H-pyrazolo[3,4-c]pyridine-3-carboxylic Acid Ethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.