accessibility menu, dialog, popup

UserName

CAS RN 2519-10-0

CAS RN 2519-10-0

IUPAC Name: (2,3,4,5-tetraphenylcyclopenta-2,4-dien-1-yl)benzene
Synonyms: (2,3,4,5-tetraphenylcyclopenta-2,4-dien-1-yl)benzene
Molecular Weight (g/mol): 446.59
Molecular Formula: C35H26
InChi Key: YGLVWOUNCXBPJF-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)C1C(=C(C(=C1C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results