CAS RN 25194-01-8
IUPAC Name:
2,6-dichloro-4-nitropyridine
Synonyms:
2,6-dichloro-4-nitropyridine
Molecular Weight (g/mol):
192.98
Molecular Formula:
C5H2Cl2N2O2
InChi Key:
BZYQSSVTQJTUDD-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CC(Cl)=NC(Cl)=C1
1
–
1
of
1
results
1
2,6-Dichloro-4-nitropyridine, 97%, Thermo Scientific Chemicals
CAS: 25194-01-8 Molecular Formula: C5H2Cl2N2O2 Molecular Weight (g/mol): 192.983 MDL Number: MFCD05670545 InChI Key: BZYQSSVTQJTUDD-UHFFFAOYSA-N Synonym: 2,6-dichloro-4-nitro-pyridine,pyridine, 2,6-dichloro-4-nitro,2 6-dichloro-4-nitropyridine,ksc555c7j,4-nitro-2,6-dichloro-pyridine,pyridine,2,6-dichloro-4-nitro,2,6-dichloro-4-nitropyridine PubChem CID: 3841070 IUPAC Name: 2,6-dichloro-4-nitropyridine SMILES: C1=C(C=C(N=C1Cl)Cl)[N+](=O)[O-]