accessibility menu, dialog, popup

UserName

CAS RN 25194-52-9

IUPAC Name: 6-bromopyridine-2-carboxamide
Synonyms: 6-bromopyridine-2-carboxamide
Molecular Weight (g/mol): 201.02
Molecular Formula: C6H5BrN2O
InChi Key: HWRUOHMDYQEWOD-UHFFFAOYSA-N
SMILES: NC(=O)C1=NC(Br)=CC=C1
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results