accessibility menu, dialog, popup

UserName

CH3SSNOH2N

CAS RN 251992-66-2

IUPAC Name: 4-[(4-methylphenyl)sulfanyl]thieno[2,3-c]pyridine-2-carboxamide
Synonyms: 4-[(4-methylphenyl)sulfanyl]thieno[2,3-c]pyridine-2-carboxamide
Molecular Weight (g/mol): 300.39
Molecular Formula: C15H12N2OS2
InChi Key: QQGWEXFLMJGCAL-UHFFFAOYSA-N
SMILES: CC1=CC=C(SC2=C3C=C(SC3=CN=C2)C(N)=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results