CAS RN 251994-14-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
ICAM-1-IN-1, MedChemExpress
MedChemExpress ICAM-1-IN-1 is a potent and selective inhibitor of E-selectin and ICAM-1 with IC50 values of 7 and 5 nM, respectively.