CAS RN 252186-79-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
3-(2,3-Dichlorobenzamido) Lamotrigine, TRC
CAS: 252186-79-1 Molecular Formula: C16 H9 Cl4 N5 O Molecular Weight (g/mol): 429.09 Synonym: N-[5-amino-6-(2,3-dichlorophenyl)-1,2,4-triazin-3-yl]-2,3-dichlorobenzamide,Lamotrigine RC D (USP),Lamotrigine Imp. F (EP) IUPAC Name: N-[5-amino-6-(2,3-dichlorophenyl)-1,2,4-triazin-3-yl]-2,3-dichlorobenzamide SMILES: Nc1nc(NC(=O)c2cccc(Cl)c2Cl)nnc1c3cccc(Cl)c3Cl
2
N-[5-Amino-6-(2,3-dichlorophenyl)-1,2,4-triazin-3-yl]-2,3-dichlorobenzamide, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.