accessibility menu, dialog, popup

UserName

CH3OPClSOH3C

CAS RN 2524-03-0

IUPAC Name: O,O-dimethyl chlorophosphonothioate
Synonyms: O,O-dimethyl chlorophosphonothioate
Molecular Weight (g/mol): 160.55
Molecular Formula: C2H6ClO2PS
InChi Key: XFBJRFNXPUCPKU-UHFFFAOYSA-N
SMILES: COP(Cl)(=S)OC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results