CAS RN 252742-72-6
IUPAC Name:
5-(chloromethyl)-2,3-dihydro-1H-1,2,4-triazol-3-one
Synonyms:
5-(chloromethyl)-1,2-dihydro-1,2,4-triazol-3-one
Molecular Weight (g/mol):
133.54
Molecular Formula:
C3H4ClN3O
InChi Key:
ZLRBJVJEQXBAAI-UHFFFAOYSA-N
SMILES:
ClCC1=NC(=O)NN1
1
–
1
of
1
results
1
5-Chloromethyl-2H-1,2,4-triazolin-3-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.