accessibility menu, dialog, popup

UserName

H3CSH

CAS RN 25276-70-4

IUPAC Name: pentadecane-1-thiol
Synonyms: pentadecane-1-thiol
Molecular Weight (g/mol): 244.48
Molecular Formula: C15H32S
InChi Key: IGMQODZGDORXEN-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCS
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results