accessibility menu, dialog, popup

UserName

CaOOOO

CAS RN 2528-16-7

IUPAC Name: calcium 2-[(benzyloxy)carbonyl]benzoate
Synonyms: calcium 2-[(benzyloxy)carbonyl]benzoate
Molecular Weight (g/mol): 295.33
Molecular Formula: C15H11CaO4
InChi Key: HLBMYHRBXPVVCC-UHFFFAOYSA-M
SMILES: [Ca++].[O-]C(=O)C1=CC=CC=C1C(=O)OCC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results