accessibility menu, dialog, popup

UserName

H3CClO

CAS RN 2528-61-2

IUPAC Name: heptanoyl chloride
Synonyms: heptanoyl chloride
Molecular Weight (g/mol): 148.63
Molecular Formula: C7H13ClO
InChi Key: UCVODTZQZHMTPN-UHFFFAOYSA-N
SMILES: CCCCCCC(Cl)=O
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)