accessibility menu, dialog, popup

UserName

CH3NNNH2N

CAS RN 252928-74-8

IUPAC Name: 3-(4-methyl-4H-1,2,4-triazol-3-yl)aniline
Synonyms: 3-(4-methyl-1,2,4-triazol-3-yl)aniline
Molecular Weight (g/mol): 174.21
Molecular Formula: C9H10N4
InChi Key: YSSZZGGTNQQAPU-UHFFFAOYSA-N
SMILES: CN1C=NN=C1C1=CC(N)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results