CAS RN 252977-51-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
TPA 023, MedChemExpress
MedChemExpress TPA 023 is a GABAA α2/α3 subtype-selective agonist, with Ki of 0.19-0.41 nM.