CAS RN 252979-56-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Banoxantrone dihydrochloride, MedChemExpress
MedChemExpress Banoxantrone dihydrochloride is a novel bioreductive agent that can be reduced to a stable, DNA-affinic compound AQ4, which is a potent topoisomerase II inhibitor.
2
Banoxantrone dihydrochloride
Prodrug topoisomerase II inhibitor