CAS RN 253-72-5
IUPAC Name:
1,6-naphthyridine
Synonyms:
1,6-naphthyridine 1,6-pyridopyridine
Molecular Weight (g/mol):
130.15
Molecular Formula:
C8H6N2
InChi Key:
VSOSXKMEQPYESP-UHFFFAOYSA-N
SMILES:
C1=CN=C2C=CN=CC2=C1