CAS RN 25339-99-5
IUPAC Name:
(2R,3R,4S,5S,6R)-2-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol; dodecanoic acid
Synonyms:
lauric acid; sucrose
Molecular Weight (g/mol):
542.62
Molecular Formula:
C24H46O13
InChi Key:
PVVVEHXCVQRLOC-AKSHDPDZSA-N
SMILES:
CCCCCCCCCCCC(O)=O.OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
1
–
3
of
3
results
1
Sucrose Monolaurate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Sucrose Monolaurate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Sucrose Monolaurate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.