CAS RN 253431-35-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
28-Epirapamycin, TRC
CAS: 253431-35-5 Molecular Formula: C51H79NO13 Molecular Weight (g/mol): 914.17 Synonym: 28-epi-Rapamycin,(31S)-Rapamycin SMILES: [H][C@]1(C(O[C@@]([H])(CC([C@H](C)/C=C([C@H](O)[C@H]2OC)\C)=O)[C@@H](C[C@@]3([H])C[C@@H](OC)[C@H](O)CC3)C)=O)N(C(C([C@@]4(O)[C@H](C)CC[C@@]([H])(C[C@H](OC)/C(C)=C/C=C/C=C/[C@@H](C)C[C@@H](C)C2=O)O4)=O)=O)CCCC1