CAS RN 25348-64-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Conduritol B, TRC
CAS: 25348-64-5 Molecular Formula: C6 H10 O4 Molecular Weight (g/mol): 146.14 IUPAC Name: (1R,2S,3S,4R)-cyclohex-5-ene-1,2,3,4-tetrol SMILES: O[C@@H]1C=C[C@@H](O)[C@H](O)[C@H]1O