accessibility menu, dialog, popup

UserName

CH3NH2NS

CAS RN 2536-91-6

IUPAC Name: 6-methyl-1,3-benzothiazol-2-amine
Synonyms: 2-benzothiazolamine, 6-methyl-
Molecular Weight (g/mol): 164.23
Molecular Formula: C8H8N2S
InChi Key: DZWTXWPRWRLHIL-UHFFFAOYSA-N
SMILES: CC1=CC=C2N=C(N)SC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results