CAS RN 25379-88-8
IUPAC Name:
methyl 2-(3,4-dihydroxyphenyl)acetate
Synonyms:
methyl 2-(3,4-dihydroxyphenyl)acetate
Molecular Weight (g/mol):
182.18
Molecular Formula:
C9H10O4
InChi Key:
UGFILLIGHGZLHE-UHFFFAOYSA-N
SMILES:
COC(=O)CC1=CC(O)=C(O)C=C1
1
–
2
of
2
results
1
3,4-Dihydroxyphenylacetic Acid Methyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
3,4-Dihydroxyphenylacetic Acid Methyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.