CAS RN 25383-06-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
[(Z)-Prop-1-enyl]phosphonic Acid (Technical Grade), TRC
CAS: 25383-06-6 Molecular Formula: C3H7O3P Molecular Weight (g/mol): 122.06 Synonym: Phosphonic acid, P-(1Z)-1-propen-1-yl- (ACI),P-(1Z)-1-Propen-1-ylphosphonic acid (ACI),Phosphonic acid, (1Z)-1-propenyl- (9CI),Phosphonic acid, 1-propenyl-, (Z)- (ZCI),Phosphonic acid, propenyl-, (Z)- (8CI),(cis-1-Propenyl)phosphonic acid,[(Z)-1-Propenyl]phosphonic acid,cis-Propenylphosphonic acid IUPAC Name: [(Z)-prop-1-enyl]phosphonic acid SMILES: C\C=C/P(=O)(O)O