CAS RN 25394-57-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Spilanthol (Technical Grade), TRC
CAS: 25394-57-4 Molecular Formula: C14 H23 N O Molecular Weight (g/mol): 221.34 Synonym: (2E,6Z,8E)-N-(2-Methylpropyl)-2,6,8-decatrienamide,(E,E,Z)-N-(2-Methylpropyl)-2,6,8-decatrienamide,(E,E,Z)- N-Isobutyl-2,6,8-decatrienamide,Affinin,2E,6Z,8E-Decatrienoic Acid N-Isobutylamide,N-Isobutyldeca-trans-2,cis-6,trans-8-trienamide; IUPAC Name: (2E,6E,8Z)-N-(2-methylpropyl)deca-2,6,8-trienamide SMILES: C\C=C/C=C/CC\C=C\C(=O)NCC(C)C