CAS RN 2540-82-1
IUPAC Name:
O,O-dimethyl ({[formyl(methyl)carbamoyl]methyl}sulfanyl)phosphonothioate
Synonyms:
formothion
Molecular Weight (g/mol):
257.26
Molecular Formula:
C6H12NO4PS2
InChi Key:
AIKKULXCBHRFOS-UHFFFAOYSA-N
SMILES:
COP(=S)(OC)SCC(=O)N(C)C=O
1
–
1
of
1
results
1
Formothion 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality