CAS RN 25405-62-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
o-Nitrophenyl Beta-D-Galactopyranoside-6-phosphate, Cyclohexylammonium Salt,, TRC
CAS: 25405-62-3 Molecular Formula: C24 H36 N O11 P Molecular Weight (g/mol): 545.52 IUPAC Name: dicyclohexyl [(2R,3R,4S,5R,6S)-3,4,5-trihydroxy-6-(2-nitrophenoxy)oxan-2-yl]methyl phosphate SMILES: O[C@H]1[C@@H](O)[C@@H](COP(=O)(OC2CCCCC2)OC3CCCCC3)O[C@@H](Oc4ccccc4[N+](=O)[O-])[C@@H]1O