accessibility menu, dialog, popup

UserName

H2NOOONH2O

CAS RN 25451-15-4

IUPAC Name: 3-(carbamoyloxy)-2-phenylpropyl carbamate
Synonyms: felbamate felbamato felbamatum 非尔氨酯 taloxa felbamyl felbatol
Molecular Weight (g/mol): 238.24
Molecular Formula: C11H14N2O4
InChi Key: WKGXYQFOCVYPAC-UHFFFAOYSA-N
SMILES: NC(=O)OCC(COC(N)=O)C1=CC=CC=C1
Percent Purity 
  • (3)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Color 
  • (4)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (2)
  • (2)
  • (3)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (4)
  • (4)