CAS RN 25462-23-1
IUPAC Name:
2-pentylpropane-1,3-diol
Synonyms:
1,3-propanediol, 2-pentyl-
Molecular Weight (g/mol):
146.23
Molecular Formula:
C8H18O2
InChi Key:
RHYUFGNCUXTFTC-UHFFFAOYSA-N
SMILES:
CCCCCC(CO)CO
1
–
1
of
1
results
1
2-n-Pentylpropane-1,3-diol, 97%, Thermo Scientific Chemicals
CAS: 25462-23-1 Molecular Formula: C8H18O2 Molecular Weight (g/mol): 146.23 MDL Number: MFCD00801123 InChI Key: RHYUFGNCUXTFTC-UHFFFAOYSA-N Synonym: 2-pentyl-1,3-propanediol,1,3-propanediol, 2-pentyl,2-n-pentylpropane-1,3-diol,2-pentylpropane-1,3diol,2-n-pentylpropane-1,3diol,2-n-pentylpropan-1,3-diol,2-n-pentyl-1,3-propanediol,2-n-pentylpropane-1, 3-diol PubChem CID: 117509 IUPAC Name: 2-pentylpropane-1,3-diol SMILES: CCCCCC(CO)CO