CAS RN 254973-02-9
IUPAC Name:
N-(7-sulfanylidene-3,7-dihydro-2,1,3-benzoxadiazol-4-yl)acetamide
Synonyms:
N-(7-sulfanylidene-3H-2,1,3-benzoxadiazol-4-yl)acetamide
Molecular Weight (g/mol):
209.22
Molecular Formula:
C8H7N3O2S
InChi Key:
JPARFDNKICYVCE-UHFFFAOYSA-N
SMILES:
CC(=O)NC1=C2NON=C2C(=S)C=C1
1
–
3
of
3
results
1
AABD-SH (4-Acetamido-7-mercapto-2,1,3-benzoxadiazole, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
AABD-SH (4-Acetamido-7-mercapto-2,1,3-benzoxadiazole, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
AABD-SH (4-Acetamido-7-mercapto-2,1,3-benzoxadiazole, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.