CAS RN 25526-93-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3’-Deoxy-3’-fluoro Thymidine, TRC
CAS: 25526-93-6 Molecular Formula: C10H13FN2O4 Molecular Weight (g/mol): 244.22 SMILES: CC1=CN([C@H]2C[C@H](F)[C@@H](CO)O2)C(=O)NC1=O