accessibility menu, dialog, popup

UserName

CH3OOFFF

CAS RN 2557-13-3

IUPAC Name: methyl 3-(trifluoromethyl)benzoate
Synonyms: methyl 3-(trifluoromethyl)benzoate
Molecular Weight (g/mol): 204.15
Molecular Formula: C9H7F3O2
InChi Key: QQHNNQCWKYFNAC-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC(=C1)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results