CAS RN 25573-43-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Eseridine, TRC
CAS: 25573-43-7 Molecular Formula: C15 H21 N3 O3 Molecular Weight (g/mol): 291.35 Synonym: 1,2-Oxazino[6,5-b]indol-6-ol, 2,3,4,4a,9,9a-hexahydro-2,4a,9-trimethyl-, methylcarbamate (ester), (4aS,9aS)- (9CI),1,2-Oxazino[6,5-b]indol-6-ol, 2,3,4,4a,9,9a-hexahydro-2,4a,9-trimethyl-, methylcarbamate (ester), (4aS-cis)-,Geneserine (6CI,7CI,8CI),(-)-Geneserine,Eseridine,Eserine aminoxide,Eserine oxide,NSC 340071,Physostigmine aminoxide IUPAC Name: [(4aS,9aS)-2,4a,9-trimethyl-4,9a-dihydro-3H-oxazino[6,5-b]indol-6-yl] N-methylcarbamate SMILES: CNC(=O)Oc1ccc2N(C)[C@H]3ON(C)CC[C@@]3(C)c2c1