CAS RN 25574-11-2
IUPAC Name:
3-(4-bromophenyl)propan-1-ol
Synonyms:
3-(4-bromophenyl)propan-1-ol
Molecular Weight (g/mol):
215.09
Molecular Formula:
C9H11BrO
InChi Key:
WODKXGCVVOOEIJ-UHFFFAOYSA-N
SMILES:
OCCCC1=CC=C(Br)C=C1
1
–
1
of
1
results
1
3-(4-Bromophenyl)-1-propanol, 98%
CAS: 25574-11-2 Molecular Formula: C9H11BrO Molecular Weight (g/mol): 215.09 MDL Number: MFCD09028724 InChI Key: WODKXGCVVOOEIJ-UHFFFAOYSA-N Synonym: 3-4-bromophenyl propan-1-ol,4-bromobenzenepropanol,3-4-bromo-phenyl-propan-1-ol,benzenepropanol, 4-bromo,3-4-bromophenyl-1-propanol,pubchem3984,3-4-bromophenyl propanol,3-4-bromophenyl-propanol,amth183,3-4-bromophenyl propyl alcohol PubChem CID: 10560614 IUPAC Name: 3-(4-bromophenyl)propan-1-ol SMILES: C1=CC(=CC=C1CCCO)Br