CAS RN 25597-16-4
IUPAC Name:
ethyl (2E)-4,4,4-trifluorobut-2-enoate
Synonyms:
ethyl (2E)-4,4,4-trifluorobut-2-enoate
Molecular Weight (g/mol):
168.12
Molecular Formula:
C6H7F3O2
InChi Key:
ZKRJCMKLCDWROR-ONEGZZNKSA-N
SMILES:
CCOC(=O)\C=C\C(F)(F)F
1
–
2
of
2
results
1
Ethyl 4,4,4-trifluorocrotonate, 98%
CAS: 25597-16-4 Molecular Formula: C6H7F3O2 Molecular Weight (g/mol): 168.12 MDL Number: MFCD00009903 InChI Key: ZKRJCMKLCDWROR-ONEGZZNKSA-N Synonym: ethyl 4,4,4-trifluorocrotonate,ethyl 2e-4,4,4-trifluorobut-2-enoate,ethyl4,4,4-trifluorocrotonate,e-ethyl 4,4,4-trifluorobut-2-enoate,4,4,4-trifluorocrotonic acid ethyl ester,ethyl e-4,4,4-trifluorobut-2-enoate,ethyl-4,4,4-trifluorocrotonate,ethyl 4,4,4-trifluorobut-2-enoate,4,4,4-trifluoro-but-2-enoic acid ethyl ester,2-butenoic acid, 4,4,4-trifluoro-, ethyl ester, 2e PubChem CID: 5371261 IUPAC Name: ethyl (E)-4,4,4-trifluorobut-2-enoate SMILES: CCOC(=O)\C=C\C(F)(F)F