CAS RN 25612-46-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Lysino Norleucine, TRC
CAS: 25612-46-8 Molecular Formula: C12 H25 N3 O4 Molecular Weight (g/mol): 275.34 IUPAC Name: (2S)-2-amino-6-[[(5S)-5-amino-5-carboxypentyl]amino]hexanoic acid SMILES: N[C@@H](CCCCNCCCC[C@H](N)C(=O)O)C(=O)O