CAS RN 256412-89-2
IUPAC Name:
2-{4-[(6-chloro-1,3-benzoxazol-2-yl)oxy]phenoxy}-N-(2-fluorophenyl)-N-methylpropanamide
Synonyms:
2-{4-[(6-chloro-1,3-benzoxazol-2-yl)oxy]phenoxy}-N-(2-fluorophenyl)-N-methylpropanamide
Molecular Weight (g/mol):
440.86
Molecular Formula:
C23H18ClFN2O4
InChi Key:
ADDQHLREJDZPMT-UHFFFAOYNA-N
SMILES:
CC(OC1=CC=C(OC2=NC3=CC=C(Cl)C=C3O2)C=C1)C(=O)N(C)C1=CC=CC=C1F
1
–
5
of
5
results
1
GB 23200.121-2021 Pesticide Mixture 6 50 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
2
Metamifop 90%, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Metamifop 90%, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Metamifop 90%, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.