CAS RN 2564467-03-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ VH 101 phenol-alkylC4-amine
VHL ligand with alkyl linker and terminal amine for onward chemistry