CAS RN 256477-09-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
UK-371804, MedChemExpress
MedChemExpress UK-371804 is a urokinase-type plasminogen activator (uPA) inhibitor with a Ki of 10 nM.