accessibility menu, dialog, popup

UserName

HOOHNONHOO

CAS RN 2566-19-0

IUPAC Name: 2-(2-{[(benzyloxy)carbonyl]amino}acetamido)acetic acid
Synonyms: (2-{[(benzyloxy)carbonyl]amino}acetamido)acetic acid
Molecular Weight (g/mol): 266.25
Molecular Formula: C12H14N2O5
InChi Key: VFRCXEHNAFUTQC-UHFFFAOYSA-N
SMILES: OC(=O)CNC(=O)CNC(=O)OCC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)