CAS RN 2566-89-4
IUPAC Name:
methyl (5Z,8Z,11Z,14Z)-icosa-5,8,11,14-tetraenoate
Synonyms:
methyl (5Z,8Z,11Z,14Z)-icosa-5,8,11,14-tetraenoate
Molecular Weight (g/mol):
318.5
Molecular Formula:
C21H34O2
InChi Key:
OFIDNKMQBYGNIW-ZKWNWVNESA-N
SMILES:
CCCCC\C=C/C\C=C/C\C=C/C\C=C/CCCC(=O)OC
1
–
1
of
1
results
1
Arachidonic Acid Methyl Ester, TRC
CAS: 2566-89-4 Molecular Formula: C21 H34 O2 Molecular Weight (g/mol): 318.49 Synonym: 5,8,11,14-Eicosatetraenoic acid, methyl ester, (all-Z)- (8CI),Arachidonic acid, methyl ester (6CI,7CI),Methyl 5Z,8Z,11Z,14Z-eicosatetraenoate,Methyl all-cis-5,8,11,14-eicosatetraenoate,Methyl arachidonate,Methyl cis,cis,cis,cis-eicosa-5,8,11,14-tetraenoate IUPAC Name: methyl (5Z,8Z,11Z,14Z)-icosa-5,8,11,14-tetraenoate SMILES: CCCCC\C=C/C\C=C/C\C=C/C\C=C/CCCC(=O)OC