CAS RN 25677-40-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
2-Pentylidenecyclohexanone, TRC
CAS: 25677-40-1 Molecular Formula: C11 H18 O Molecular Weight (g/mol): 166.26 Synonym: 2-Pentylidenecyclohexanone,2-Pentylidenecyclohexanone IUPAC Name: (2E)-2-pentylidenecyclohexan-1-one SMILES: CCCC\C=C\1/CCCCC1=O
2
2-Pentylidenecyclohexanone 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality