accessibility menu, dialog, popup

UserName

CH3NNOBr

CAS RN 256956-42-0

IUPAC Name: 3-[4-(bromomethyl)phenyl]-5-methyl-1,2,4-oxadiazole
Synonyms: 3-[4-(bromomethyl)phenyl]-5-methyl-1,2,4-oxadiazole
Molecular Weight (g/mol): 253.1
Molecular Formula: C10H9BrN2O
InChi Key: BMXSEDLZMUPKSH-UHFFFAOYSA-N
SMILES: CC1=NC(=NO1)C1=CC=C(CBr)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results