CAS RN 25700-11-2
IUPAC Name:
2-(1H-pyrazol-1-yl)pyridine
Synonyms:
2-(pyrazol-1-yl)pyridine
Molecular Weight (g/mol):
145.17
Molecular Formula:
C8H7N3
InChi Key:
XXTPHXNBKRVYJI-UHFFFAOYSA-N
SMILES:
C1=CN(N=C1)C1=CC=CC=N1
1
–
1
of
1
results
1
2-(1H-Pyrazol-1-yl)pyridine, Thermo Scientific™
CAS: 25700-11-2 Molecular Formula: C8H7N3 Molecular Weight (g/mol): 145.17 MDL Number: 09033534 InChI Key: XXTPHXNBKRVYJI-UHFFFAOYSA-N IUPAC Name: 2-(1H-pyrazol-1-yl)pyridine SMILES: C1=CN(N=C1)C1=CC=CC=N1