CAS RN 25713-60-4
IUPAC Name:
tris(2,4,6-tribromophenoxy)-1,3,5-triazine
Synonyms:
tris(2,4,6-tribromophenoxy)-1,3,5-triazine
Molecular Weight (g/mol):
1067.43
Molecular Formula:
C21H6Br9N3O3
InChi Key:
BDFBPPCACYFGFA-UHFFFAOYSA-N
SMILES:
BrC1=CC(Br)=C(OC2=NC(OC3=C(Br)C=C(Br)C=C3Br)=NC(OC3=C(Br)C=C(Br)C=C3Br)=N2)C(Br)=C1
1
–
3
of
3
results
1
2,4,6-Tris(2,4,6-tribomophenoxy)-1,3,5-triazine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2,4,6-Tris(2,4,6-tribomophenoxy)-1,3,5-triazine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2,4,6-Tris(2,4,6-tribomophenoxy)-1,3,5-triazine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.