CAS RN 257292-29-8
IUPAC Name:
3-[(5E)-7-(2,4-dimethoxyphenyl)-1,4-thiazepan-5-ylidene]-6-methyl-3,4-dihydro-2H-pyran-2,4-dione
Synonyms:
3-[(5E)-7-(2,4-dimethoxyphenyl)-1,4-thiazepan-5-ylidene]-6-methylpyran-2,4-dione
Molecular Weight (g/mol):
375.44
Molecular Formula:
C19H21NO5S
InChi Key:
FJDFKOSFDPIMBV-NBVRZTHBNA-N
SMILES:
COC1=CC=C(C2C\C(NCCS2)=C2/C(=O)OC(C)=CC2=O)C(OC)=C1